ENA090493113 Enamine

N-{[4-(benzyloxy)phenyl]methyl}-2-chloro-N-ethylacetamide

  • MDL number MFCD08444118
  • Cas Number : 923777-57-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-22972

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-22972
Availability details for
Quantity
  • Availability
  • Add to Cart