ENA090493115 Enamine

2-(2-chloroacetamido)-N-(3-methylphenyl)benzamide

  • MDL number MFCD08444090
  • Cas Number : 22312-75-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-21740

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-21740
Availability details for
Quantity
  • Availability
  • Add to Cart