ENA090493116 Enamine

[1,2,4]triazolo[1,5-a]pyrimidine-2-thiol

  • MDL number MFCD08444303
  • Cas Number : 66234-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24654

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24654
Availability details for
Quantity
  • Availability
  • Add to Cart