ENA090493121 Enamine

1-cyclopropyl-5-methoxy-2-methyl-1H-indole-3-carboxylic acid

  • MDL number MFCD01566850
  • Cas Number : 300541-60-0
  • Molecular Weight : 245.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24659

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24659
Availability details for
Quantity
  • Availability
  • Add to Cart