ENA090493135 Enamine

(2E)-3-(2-cyanophenyl)acrylic acid

  • MDL number MFCD00189382
  • Cas Number : 61147-65-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 255...257
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24714

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24714
Availability details for
Quantity
  • Availability
  • Add to Cart