ENA090493156 Enamine

5-thien-2-ylisoxazole-3-carboxylic acid

  • MDL number MFCD04113955
  • Cas Number : 763109-71-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173...175
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24764

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24764
Availability details for
Quantity
  • Availability
  • Add to Cart