ENA090493159 Enamine

2-chloro-N-[2-(2-methylphenyl)ethyl]acetamide

  • MDL number MFCD08444137
  • Cas Number : 141463-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-23170

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23170
Availability details for
Quantity
  • Availability
  • Add to Cart