ENA090493161 Enamine

6-(2,5-dimethylthien-3-yl)-3-methylisoxazolo[5,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD08166694
  • Cas Number : 900137-00-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 256...258
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24770

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24770
Availability details for
Quantity
  • Availability
  • Add to Cart