ENA090493168 Enamine

2-chloro-N-[4-methyl-3-(morpholine-4-carbonyl)-5-phenylthiophen-2-yl]acetamide

  • MDL number MFCD08444139
  • Cas Number : 924376-11-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-23190

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23190
Availability details for
Quantity
  • Availability
  • Add to Cart