ENA090493177 Enamine

3-(2-chloroacetamido)-N-ethylbenzamide

  • MDL number MFCD08444144
  • Cas Number : 923243-30-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23207

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23207
Availability details for
Quantity
  • Availability
  • Add to Cart