ENA090493179 Enamine

2-chloro-N-[4-methyl-3-(methylsulfamoyl)phenyl]acetamide

  • MDL number MFCD08444148
  • Cas Number : 923104-49-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23268

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23268
Availability details for
Quantity
  • Availability
  • Add to Cart