ENA090493203 Enamine

1-[(4-chlorophenyl)methyl]-1,4-diazepane

  • MDL number MFCD03407501
  • Cas Number : 40389-65-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-25345

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25345
Availability details for
Quantity
  • Availability
  • Add to Cart