ENA090493209 Enamine

5-(chloromethyl)-3-phenyl-1,2-oxazole

  • MDL number MFCD01935949
  • Cas Number : 1011-37-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69-73
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-25358

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25358
Availability details for
Quantity
  • Availability
  • Add to Cart