ENA090493225 Enamine

methyl 3-(2-oxo-2H-1,4-benzoxazin-3-yl)propanoate

  • MDL number MFCD06251781
  • Cas Number : 852538-97-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25379

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25379
Availability details for
Quantity
  • Availability
  • Add to Cart