ENA090493233 Enamine

6-(furan-2-yl)-1-(propan-2-yl)-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD08166702
  • Cas Number : 900137-06-6
  • Molecular Weight : 271.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 276-279
Iupac Name 6-(2-furyl)-1-isopropyl-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid
Inchi Code 1S/C14H13N3O3/c1-8(2)17-13-10(7-15-17)9(14(18)19)6-11(16-13)12-4-3-5-20-12/h3-8H,1-2H3,(H,18,19)
Inchi Key LVECVVOZMOWPNV-UHFFFAOYSA-N

Description

Melting Point 276-279
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-25390

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-25390
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart