ENA090493254 Enamine

2-chloro-N-[4-(3,4-dichlorophenyl)-1,3-thiazol-2-yl]-N-(prop-2-en-1-yl)acetamide

  • MDL number MFCD08444108
  • Cas Number : 924133-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-22718

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-22718
Availability details for
Quantity
  • Availability
  • Add to Cart