ENA090493255 Enamine

2-(piperazin-1-ylmethyl)quinazolin-4(3H)-one

  • MDL number MFCD05022545
  • Cas Number : 725690-23-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 167...169
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25424

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25424
Availability details for
Quantity
  • Availability
  • Add to Cart