ENA090493273 Enamine

4-[(furan-2-yl)methyl]-5-(propan-2-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD06758265
  • Cas Number : 777875-15-7
  • Molecular Weight : 223.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-25453

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25453
Availability details for
Quantity
  • Availability
  • Add to Cart