ENA090493274 Enamine

5-isopropyl-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD06803595
  • Cas Number : 1711-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25457

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-25457
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart