ENA090493289 Enamine

4-[(4-methyl-4H-1,2,4-triazol-3-yl)sulfanyl]aniline

  • MDL number MFCD08444383
  • Cas Number : 497851-88-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148-149
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25479

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25479
Availability details for
Quantity
  • Availability
  • Add to Cart