ENA090493291 Enamine

5-iodo-6-nitro-1,3-dioxaindane

  • MDL number MFCD00188400
  • Cas Number : 7106-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60-61
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24502

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24502
Availability details for
Quantity
  • Availability
  • Add to Cart