ENA090493293 Enamine

methyl bromo(4-fluorophenyl)acetate

  • MDL number MFCD08444284
  • Cas Number : 71783-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24514

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24514
Availability details for
Quantity
  • Availability
  • Add to Cart