ENA090493295 Enamine

(1E)-1-[1-(difluoromethyl)-1H-benzimidazol-2-yl]ethanone oxime

  • MDL number MFCD08166737
  • Cas Number : 1955564-13-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25488

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25488
Availability details for
Quantity
  • Availability
  • Add to Cart