ENA090493313 Enamine

2-(2-chloroacetamido)-N-(4-fluorophenyl)benzamide

  • MDL number MFCD08444089
  • Cas Number : 923843-56-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-18524

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-18524
Availability details for
Quantity
  • Availability
  • Add to Cart