ENA100800951 Enamine

1-[(2,2,2-trifluoroethoxy)carbonyl]piperidine-4-carboxylic acid

  • MDL number MFCD08444270
  • Cas Number : 923249-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24485

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24485
Availability details for
Quantity
  • Availability
  • Add to Cart