ENA100800959 Enamine

1-(2-chloroacetyl)-N-phenylpiperidine-4-carboxamide

  • MDL number MFCD08444301
  • Cas Number : 923171-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-24638

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24638
Availability details for
Quantity
  • Availability
  • Add to Cart