ENA100800960 Enamine

2-chloro-1-{4-[(3,5-dimethyl-1,2-oxazol-4-yl)sulfonyl]piperazin-1-yl}ethan-1-one

  • MDL number MFCD08691113
  • Cas Number : 923203-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24639

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24639
Availability details for
Quantity
  • Availability
  • Add to Cart