ENA100800961 Enamine

2-chloro-N-{1-[(2-chlorophenyl)methyl]-1H-pyrazol-5-yl}acetamide

  • MDL number MFCD08444306
  • Cas Number : 956769-09-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24679

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24679
Availability details for
Quantity
  • Availability
  • Add to Cart