ENA100800970 Enamine

5-cyclopropaneamido-3-methylthiophene-2-carboxylic acid

  • MDL number MFCD08444394
  • Cas Number : 923126-67-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25515

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25515
Availability details for
Quantity
  • Availability
  • Add to Cart