ENA100800972 Enamine

3-(1-methylidene-3-oxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid

  • MDL number MFCD08444397
  • Cas Number : 923120-68-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-25520

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25520
Availability details for
Quantity
  • Availability
  • Add to Cart