ENA100800977 Enamine

4-[1-(difluoromethyl)-1H-1,3-benzodiazol-2-yl]aniline

  • MDL number MFCD08444403
  • Cas Number : 924118-90-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25534

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25534
Availability details for
Quantity
  • Availability
  • Add to Cart