ENA100800981 Enamine

6-phenyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylic acid

  • MDL number MFCD08444411
  • Cas Number : 923792-95-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25551
Availability details for
Quantity
  • Availability
  • Add to Cart