ENA100800982 Enamine

2-chloro-N-(4-{5H,6H,7H,8H,9H-[1,2,4]triazolo[4,3-a]azepin-3-yl}phenyl)acetamide

  • MDL number MFCD08444412
  • Cas Number : 923696-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-25552

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25552
Availability details for
Quantity
  • Availability
  • Add to Cart