ENA100800984 Enamine

1-[(furan-2-yl)methyl]-1H-1,2,3,4-tetrazole-5-thiol

  • MDL number MFCD08444413
  • Cas Number : 204070-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25557

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25557
Availability details for
Quantity
  • Availability
  • Add to Cart