ENA100800985 Enamine

[2-(thiophen-2-yl)ethyl]thiourea

  • MDL number MFCD08444414
  • Cas Number : 923255-84-9
  • Molecular Weight : 186.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25559

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25559
Availability details for
Quantity
  • Availability
  • Add to Cart