ENA100800993 Enamine

1-(2-chloroacetyl)-2,2,6,6-tetramethylpiperidin-4-one

  • MDL number MFCD08444423
  • Cas Number : 923177-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-25579

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25579
Availability details for
Quantity
  • Availability
  • Add to Cart