ENA100800994 Enamine

2-(1-methylidene-3-oxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid

  • MDL number MFCD08444424
  • Cas Number : 62100-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-25581

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25581
Availability details for
Quantity
  • Availability
  • Add to Cart