ENA100800996 Enamine

2-(quinolin-8-yl)acetonitrile

  • MDL number MFCD08444426
  • Cas Number : 66819-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25587

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25587
Availability details for
Quantity
  • Availability
  • Add to Cart