ENA100801000 Enamine

2-chloro-N-[3-cyano-1-(4-methoxyphenyl)-4,5-dimethyl-1H-pyrrol-2-yl]acetamide

  • MDL number MFCD08444430
  • Cas Number : 923870-33-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25602

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25602
Availability details for
Quantity
  • Availability
  • Add to Cart