ENA100801007 Enamine

5-(4-fluorophenyl)-1,3-oxazole-2-thiol

  • MDL number MFCD08444441
  • Cas Number : 923218-69-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25632

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25632
Availability details for
Quantity
  • Availability
  • Add to Cart