ENA100801018 Enamine

3-methyl-6-[2-(trifluoromethyl)phenyl]-[1,2]oxazolo[5,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD08444454
  • Cas Number : 924383-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25660

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25660
Availability details for
Quantity
  • Availability
  • Add to Cart