ENA100801026 Enamine

2-phenyl-2-(piperazin-1-yl)acetamide

  • MDL number MFCD08444472
  • Cas Number : 923163-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25689

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25689
Availability details for
Quantity
  • Availability
  • Add to Cart