ENA100801027 Enamine

5-(furan-2-amido)-3-methylthiophene-2-carboxylic acid

  • MDL number MFCD08444473
  • Cas Number : 923826-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25692

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25692
Availability details for
Quantity
  • Availability
  • Add to Cart