ENA100801031 Enamine

4-(2-furylmethyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD08444481
  • Cas Number : 660417-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25712

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25712
Availability details for
Quantity
  • Availability
  • Add to Cart