ENA100801053 Enamine

N-cyclopropyl-2-(piperazin-1-yl)propanamide

  • MDL number MFCD08691122
  • Cas Number : 923225-24-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25871

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25871
Availability details for
Quantity
  • Availability
  • Add to Cart