ENA100801061 Enamine

5-chloro-1-[(4-fluorophenyl)methyl]-3-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD08691128
  • Cas Number : 956190-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25896

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25896
Availability details for
Quantity
  • Availability
  • Add to Cart