ENA100801064 Enamine

1-(4-amino-2-chlorophenyl)pyrrolidin-2-one

  • MDL number MFCD08691131
  • Cas Number : 444002-88-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184...186
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25903

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25903
Availability details for
Quantity
  • Availability
  • Add to Cart