ENA100801067 Enamine

2-[4-(2-chloroacetyl)piperazin-1-yl]-2-(4-fluorophenyl)acetonitrile

  • MDL number MFCD08691134
  • Cas Number : 923230-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25908

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25908
Availability details for
Quantity
  • Availability
  • Add to Cart