ENA100801070 Enamine

2-chloro-N-(3-cyano-4,5-dimethyl-1-phenyl-1H-pyrrol-2-yl)acetamide

  • MDL number MFCD08691137
  • Cas Number : 923842-98-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25913

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25913
Availability details for
Quantity
  • Availability
  • Add to Cart