ENA100801072 Enamine

4-[(furan-2-yl)methyl]-5-(morpholin-4-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD08691140
  • Cas Number : 923789-06-4
  • Molecular Weight : 266.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 127-128
Iupac Name 4-(2-furylmethyl)-5-(4-morpholinyl)-2,4-dihydro-3H-1,2,4-triazole-3-thione
Inchi Code 1S/C11H14N4O2S/c18-11-13-12-10(14-3-6-16-7-4-14)15(11)8-9-2-1-5-17-9/h1-2,5H,3-4,6-8H2,(H,13,18)
Inchi Key CODKMDZDCCCUJI-UHFFFAOYSA-N

Description

Melting Point 127-128
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-25916

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25916
Availability details for
Quantity
  • Availability
  • Add to Cart